Assay Details
| Gene Symbol: Tmem54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034151.1 | NP_001029323.1 | 3 - 4 | 397 | 70 | |
| GenBank mRNA | BC101399.1 | - | 3 - 4 | 397 | 7933358 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem54
Genomic Map