Assay Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107978.2 | NP_001101448.1 | 2 - 3 | 196 | 85 | |
| GenBank mRNA | BC168964.1 | - | 2 - 3 | 196 | 7130611 | 85 |
| FQ226285.1 | - | 2 - 3 | 208 | 85 | ||
| FQ231750.1 | - | 2 - 3 | 217 | 85 | ||
| FQ233930.1 | - | 2 - 3 | 208 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Utp11l
Genomic Map