Assay Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107978.2 | NP_001101448.1 | 4 - 5 | 400 | 102 | |
| GenBank mRNA | BC168964.1 | - | 4 - 5 | 400 | 7130611 | 102 |
| FQ226285.1 | - | 4 - 5 | 412 | 102 | ||
| FQ231750.1 | - | 4 - 5 | 421 | 102 | ||
| FQ233930.1 | - | 4 - 5 | 412 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Utp11l
Genomic Map