Assay Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107972.1 | NP_001101442.1 | 5 - 6 | 6023 | 61 | |
| XM_006238779.2 | XP_006238841.1 | 6 - 7 | 9175 | 61 | ||
| XM_008764014.1 | XP_008762236.1 | 5 - 6 | 9272 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164255 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99234 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164260 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164255 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164255 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164260 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164260 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164255 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480771 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93800 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17683 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871620 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871619 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871616 | - | - |
| Rat Genome 230 2.0 Array | 1393703_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393703_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500009489.rn.1 | TC0500001361.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500009487.rn.1 | TC0500001361.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393703_PM_at | - | - |
| Rat Expression Set 230 | 1393703_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21279704 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363842 | - | - |

Hivep3
Genomic Map