Assay Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750036.3 | XP_003750084.3 | 6 - 7 | 849 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb8os| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750036.3 | XP_003750084.3 | 6 - 7 | 849 | 147 | |
Set Membership: |
|