Assay Details
| Gene Symbol: RGD1308878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134522.1 | NP_001127994.1 | 2 - 3 | 454 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308878| Gene Symbol: RGD1308878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134522.1 | NP_001127994.1 | 2 - 3 | 454 | 60 | |
Set Membership: |
|