Assay Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048185.1 | NP_001041650.1 | 4 - 5 | 740 | 82 | |
| XM_008764233.1 | XP_008762455.1 | 4 - 5 | 932 | 82 | ||
| GenBank mRNA | BC091231.1 | - | 4 - 5 | 724 | 82 | |
| BC105628.1 | - | 4 - 5 | 740 | 7322772 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agmat
Genomic Map