Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113780.1 | NP_001107252.1 | 3 - 4 | 325 | 59 | |
| GenBank mRNA | BC158875.1 | - | 3 - 4 | 325 | 7934037 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem222
Genomic Map