Assay Details
| Gene Symbol: Trit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108676.1 | NP_001102146.1 | 4 - 5 | 616 | 73 | |
| XM_006238804.2 | XP_006238866.1 | 4 - 5 | 1155 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trit1
Genomic Map