Assay Details
| Gene Symbol: Pla2g5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239139.2 | XP_006239201.1 | 1 - 2 | 309 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239139.2 transcripts.
Set Membership: |
|

Pla2g5
Genomic Map