Assay Details
| Gene Symbol: Fam76a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108686.1 | NP_001102156.1 | 6 - 7 | 837 | 86 | |
| GenBank mRNA | FQ211538.1 | - | 4 - 5 | 770 | 86 | |
| FQ222884.1 | - | 6 - 7 | 839 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam76a
Genomic Map