Assay Details
| Gene Symbol: Mfsd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106683.1 | NP_001100153.1 | 2 - 3 | 374 | 76 | |
| XM_006238792.2 | XP_006238854.1 | 2 - 3 | 354 | 76 | ||
| XM_008764001.1 | XP_008762223.1 | 2 - 3 | 351 | 76 | ||
| GenBank mRNA | BC161915.1 | - | 2 - 3 | 361 | 8369688 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd2a
Genomic Map