Assay Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011983.1 | NP_001011983.1 | 2 - 3 | 345 | 59 | |
| GenBank mRNA | BC079342.1 | - | 2 - 3 | 345 | 7106851 | 59 |
| FQ228932.1 | - | 2 - 3 | 324 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Coq6
Genomic Map