Assay Details
| Gene Symbol: Asb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011984.1 | NP_001011984.1 | 7 - 8 | 1232 | 61 | |
| XM_006240464.2 | XP_006240526.1 | 5 - 6 | 1877 | 61 | ||
| GenBank mRNA | BC085882.1 | - | 7 - 8 | 1232 | 7191983 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Asb2
Genomic Map