Assay Details
| Gene Symbol: LOC100909439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764916.1 | XP_008763138.1 | 3 - 4 | 869 | 76 | |
| Gene Symbol: Asb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011984.1 | NP_001011984.1 | 8 - 9 | 1791 | 76 | |
| XM_006240464.2 | XP_006240526.1 | 6 - 7 | 2436 | 76 | ||
| GenBank mRNA | BC085882.1 | - | 8 - 9 | 1791 | 7191983 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909439
Genomic Map