Assay Details
| Gene Symbol: Calm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FQ211760.1 | - | 1 - 2 | 185 | 76 | |
| FQ212721.1 | - | 523 | 76 | |||
| FQ213227.1 | - | 1 - 2 | 231 | 76 | ||
| FQ225380.1 | - | 1 - 2 | 144 | 76 | ||
| FQ226971.1 | - | 1 - 2 | 190 | 76 | ||
| FQ227186.1 | - | 1 - 2 | 190 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Calm1
Genomic Map