Assay Details
| Gene Symbol: Ndufb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FQ210856.1 | - | 2 - 3 | 200 | 88 | |
| FQ212372.1 | - | 2 - 3 | 200 | 88 | ||
| FQ217107.1 | - | 2 - 3 | 188 | 88 | ||
| FQ223496.1 | - | 2 - 3 | 188 | 88 | ||
| FQ224297.1 | - | 2 - 3 | 200 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufb1
Genomic Map