Assay Details
| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206846.2 | NP_996728.1 | 1 - 2 | 49 | 78 | |
| GenBank mRNA | BC166870.1 | - | 1 - 2 | 49 | 6922110 | 78 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166870.1, NM_206846.2 transcripts.
Set Membership: |
|

Ifi27l2b
Genomic Map