Assay Details
| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206846.2 | NP_996728.1 | 2 - 3 | 80 | 76 | |
| GenBank mRNA | BC166870.1 | - | 2 - 3 | 80 | 6922110 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Ifi27l2b
Genomic Map