Assay Details
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC083569.1 | - | 12 - 13 | 1671 | 61 | |
| BC098927.1 | - | 1734 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC083569.1 | - | 12 - 13 | 1671 | 61 | |
| BC098927.1 | - | 1734 | 61 | |||
Set Membership: |
|