Assay Details
| Gene Symbol: Serpina10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133617.2 | NP_598301.2 | 1 - 2 | 89 | 92 | |
| XM_008764736.1 | XP_008762958.1 | 1 - 2 | 107 | 92 | ||
| GenBank mRNA | BC088113.1 | - | 1 - 2 | 89 | 7326099 | 92 |
| Gene Symbol: LOC100909524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750227.3 | XP_003750275.1 | 1 - 2 | 107 | 92 | |
| XM_006240518.2 | XP_006240580.1 | 1 - 2 | 107 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088113.1, NM_133617.2, XM_003750227.3, XM_006240518.2, XM_008764736.1 transcripts.
Set Membership: |
|

Serpina10
Genomic Map