Assay Details
| Gene Symbol: Itpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191985.1 | NP_001178914.1 | 8 - 9 | 740 | 66 | |
| XM_008764841.1 | XP_008763063.1 | 8 - 9 | 1734 | 66 | ||
| XM_008764842.1 | XP_008763064.1 | 8 - 9 | 2205 | 66 | ||
| XM_008764843.1 | XP_008763065.1 | 8 - 9 | 4712 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itpk1
Genomic Map