Assay Details
| Gene Symbol: Aimp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053757.1 | NP_446209.1 | 3 - 4 | 295 | 85 | |
| GenBank mRNA | BC098675.1 | - | 3 - 4 | 295 | 7322792 | 85 |
| FQ226969.1 | - | 1 - 2 | 71 | 85 | ||
| FQ229959.1 | - | 1 - 2 | 71 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aimp1
Genomic Map