Assay Details
| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191984.1 | NP_001178913.1 | 4 - 5 | 581 | 60 | |
| XM_006239263.2 | XP_006239325.1 | 4 - 5 | 671 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax7
Genomic Map