Assay Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173556.1 | NP_001167027.1 | 2 - 3 | 168 | 67 | |
| GenBank mRNA | BC166932.1 | - | 2 - 3 | 139 | 6921482 | 67 |
| FM054041.1 | - | 2 - 3 | 168 | 67 | ||
| FQ221358.1 | - | 2 - 3 | 96 | 67 | ||
| FQ228668.1 | - | 2 - 3 | 170 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Minos1
Genomic Map