Assay Details
| Gene Symbol: Mmp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053606.2 | NP_446058.1 | 2 - 3 | 306 | 73 | |
| GenBank mRNA | AB010960.1 | - | 2 - 3 | 467 | 73 | |
| BC086586.1 | - | 2 - 3 | 306 | 7309721 | 73 | |
| FQ220321.1 | - | 2 - 3 | 312 | 73 | ||
| FQ220929.1 | - | 2 - 3 | 309 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp23
Genomic Map