Assay Details
| Gene Symbol: Mmp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053606.2 | NP_446058.1 | 6 - 7 | 888 | 76 | |
| GenBank mRNA | AB010960.1 | - | 6 - 7 | 1049 | 76 | |
| BC086586.1 | - | 6 - 7 | 888 | 7309721 | 76 | |
| FQ220321.1 | - | 6 - 7 | 894 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp23
Genomic Map