Assay Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017103.2 | NP_058799.2 | 8 - 9 | 2035 | 61 | |
| GenBank mRNA | BC081815.1 | - | 8 - 9 | 2035 | 7110428 | 61 |
| L46816.1 | - | 2050 | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tceb3
Genomic Map