Assay Details
| Gene Symbol: Taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115036.1 | NP_001108508.1 | 2 - 3 | 322 | 90 | |
| XM_006239067.2 | XP_006239129.1 | 3 - 4 | 441 | 90 | ||
| XM_006239068.2 | XP_006239130.1 | 2 - 3 | 426 | 90 | ||
| GenBank mRNA | BC158610.1 | - | 2 - 3 | 301 | 7374180 | 90 |
| FQ225384.1 | - | 2 - 3 | 321 | 90 | ||
| FQ232337.1 | - | 2 - 3 | 321 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf12
Genomic Map