Assay Details
| Gene Symbol: Taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115036.1 | NP_001108508.1 | 5 - 6 | 612 | 92 | |
| XM_006239067.2 | XP_006239129.1 | 6 - 7 | 731 | 92 | ||
| XM_006239068.2 | XP_006239130.1 | 5 - 6 | 716 | 92 | ||
| GenBank mRNA | BC107929.1 | - | 6 - 7 | 746 | 92 | |
| BC158610.1 | - | 5 - 6 | 591 | 7374180 | 92 | |
| FQ225384.1 | - | 5 - 6 | 611 | 92 | ||
| FQ232337.1 | - | 5 - 6 | 611 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf12
Genomic Map