Assay Details
| Gene Symbol: Ncmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253918.1 | NP_001240847.1 | 1 - 2 | 150 | 91 | |
| GenBank mRNA | CB782535.1 | - | 1 - 2 | 150 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253918.1 transcripts.
Set Membership: |
|

Ncmap
Genomic Map