Assay Details
| Gene Symbol: LOC100911993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751808.3 | XP_003751856.1 | 7 - 8 | 3705 | 71 | |
| XM_006254970.2 | XP_006255032.1 | 7 - 8 | 3710 | 71 | ||
| Gene Symbol: Usp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198785.1 | NP_942080.1 | 22 - 23 | 2906 | 71 | |
| XM_006239234.2 | XP_006239296.1 | 23 - 24 | 2940 | 71 | ||
| XM_006239235.2 | XP_006239297.1 | 23 - 24 | 2937 | 71 | ||
| XM_006239236.2 | XP_006239298.1 | 23 - 24 | 2937 | 71 | ||
| XM_006239237.2 | XP_006239299.1 | 23 - 24 | 2934 | 71 | ||
| GenBank mRNA | AB073880.1 | - | 22 - 23 | 2906 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911993
Genomic Map