Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128189.2 | NP_001121661.1 | 10 - 11 | 1248 | 105 | |
| XM_006239073.2 | XP_006239135.1 | 10 - 11 | 1218 | 105 | ||
| XM_006239074.1 | XP_006239136.1 | 10 - 11 | 1542 | 105 | ||
| XM_006239075.2 | XP_006239137.1 | 10 - 11 | 1732 | 105 | ||
| XM_006239077.2 | XP_006239139.1 | 9 - 10 | 1178 | 105 | ||
| XM_008764172.1 | XP_008762394.1 | 10 - 11 | 2886 | 105 | ||
| XM_008764173.1 | XP_008762395.1 | 11 - 12 | 1272 | 105 | ||
| GenBank mRNA | BC160834.1 | - | 10 - 11 | 1248 | 7456254 | 105 |
More Information
Updated Mapping Information
Set Membership: |
|

Rcc1
Genomic Map