Assay Details
| Gene Symbol: Ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107911.1 | NP_001101381.1 | 5 - 6 | 651 | 71 | |
| GenBank mRNA | BC162025.1 | - | 5 - 6 | 650 | 9099457 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r8
Genomic Map