Assay Details
| Gene Symbol: Ifnlr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191868.1 | NP_001178797.1 | 4 - 5 | 506 | 74 | |
| XM_006239219.2 | XP_006239281.1 | 3 - 4 | 1432 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnlr1
Genomic Map