Assay Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107979.1 | NP_001101449.1 | 1 - 2 | 50 | 74 | |
| XM_008764016.1 | XP_008762238.1 | 1 - 2 | 184 | 74 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107979.1,XM_008764016.1 transcripts.
Set Membership: |
|

Fhl3
Genomic Map