Assay Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053475.1 | NP_445927.1 | 2 - 3 | 600 | 75 | |
| GenBank mRNA | AJ007016.1 | - | 2 - 3 | 600 | 75 | |
| BC060549.1 | - | 2 - 3 | 672 | 6891377 | 75 | |
| FQ230760.1 | - | 2 - 3 | 182 | 75 | ||
| FQ231532.1 | - | 2 - 3 | 306 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptp4a2
Genomic Map