Assay Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053475.1 | NP_445927.1 | 98 | |||
| GenBank mRNA | AJ007016.1 | - | 98 | |||
| BC060549.1 | - | 6891377 | 98 | |||
| FQ230760.1 | - | 98 | ||||
| FQ231532.1 | - | 98 | ||||

Ptp4a2
Genomic Map