Assay Details
| Gene Symbol: Pabpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100538.1 | NP_001094008.1 | 6 - 7 | 1241 | 87 | |
| XM_006238798.1 | XP_006238860.1 | 6 - 7 | 1781 | 87 | ||
| XM_006238799.1 | XP_006238861.1 | 6 - 7 | 1781 | 87 | ||
| XM_006238800.1 | XP_006238862.1 | 6 - 7 | 1781 | 87 | ||
| XM_006238801.1 | XP_006238863.1 | 6 - 7 | 1781 | 87 | ||
| GenBank mRNA | BC088337.1 | - | 1 - 2 | 140 | 87 | |
| BC166452.1 | - | 6 - 7 | 1246 | 8372968 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pabpc4
Genomic Map