Assay Details
| Gene Symbol: RGD1308923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108698.1 | NP_001102168.1 | 2 - 3 | 407 | 60 | |
| XM_006239585.2 | XP_006239647.1 | 2 - 3 | 1088 | 60 | ||
| GenBank mRNA | FQ234838.1 | - | 2 - 3 | 173 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1308923
Genomic Map