Assay Details
| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109267.1 | NP_001102737.1 | 4 - 5 | 364 | 79 | |
| XM_006238968.2 | XP_006239030.1 | 4 - 5 | 384 | 79 | ||
| XM_006238969.2 | XP_006239031.1 | 3 - 4 | 320 | 79 | ||
| XM_006238970.2 | XP_006239032.1 | 2 - 3 | 199 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zcchc17
Genomic Map