Assay Details
| Gene Symbol: RGD1564482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036617.1 | 3 - 4 | 256 | 80 | ||
| GenBank mRNA | AA997706.1 | - | 1 - 2 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1564482
Genomic Map