Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127591.1 | NP_001121063.1 | 7 - 8 | 1174 | 72 | |
| XM_008764385.1 | XP_008762607.1 | 7 - 8 | 1793 | 72 | ||
| XM_008764386.1 | XP_008762608.1 | 7 - 8 | 1960 | 72 | ||
| XM_008764387.1 | XP_008762609.1 | 7 - 8 | 1347 | 72 | ||
| GenBank mRNA | BC166429.1 | - | 7 - 8 | 1174 | 7128351 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map