Assay Details
| Gene Symbol: LOC100911725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243892.2 | XP_006243954.1 | 1249 | 73 | ||
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019333.1 | NP_062206.1 | 13 - 14 | 1318 | 73 | |
| XM_006243840.2 | XP_006243902.1 | 12 - 13 | 1277 | 73 | ||
| XM_006243841.2 | XP_006243903.1 | 12 - 13 | 1271 | 73 | ||
| XM_006243842.2 | XP_006243904.1 | 12 - 13 | 1239 | 73 | ||
| XM_008766563.1 | XP_008764785.1 | 12 - 13 | 2318 | 73 | ||
| GenBank mRNA | EU404101.1 | - | 12 - 13 | 1265 | 73 | |
| EU404103.1 | - | 11 - 12 | 1381 | 73 | ||
| M64797.1 | - | 13 - 14 | 1318 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911725
Genomic Map