Assay Details
| Gene Symbol: Twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135238.1 | NP_001128710.1 | 8 - 9 | 1036 | 95 | |
| GenBank mRNA | BC158614.1 | - | 8 - 9 | 984 | 7386008 | 95 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Twf2
Genomic Map