Assay Details
| Gene Symbol: Clca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107450.1 | NP_001100920.1 | 2 - 3 | 518 | 62 | |
| XM_008761497.1 | XP_008759719.1 | 2 - 3 | 1670 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clca5
Genomic Map