Assay Details
| Gene Symbol: Pifo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001077265.5 | XP_001077265.2 | 5 - 6 | 686 | 71 | |
| XM_003749376.3 | XP_003749424.1 | 5 - 6 | 697 | 71 | ||
More Information
Set Membership: |
|

Pifo
Genomic Map