Assay Details
| Gene Symbol: LOC685067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749409.3 | XP_003749457.1 | 5 - 6 | 814 | 66 | |
| GenBank mRNA | FQ221477.1 | - | 738 | 66 | ||
| FQ222982.1 | - | 5 - 6 | 721 | 66 | ||
More Information
Set Membership: |
|

LOC685067
Genomic Map