Assay Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019359.1 | NP_062232.1 | 4 - 5 | 614 | 65 | |
| GenBank mRNA | BC062020.1 | - | 4 - 5 | 617 | 5623281 | 65 |
| FQ228936.1 | - | 1 - 2 | 51 | 65 | ||
| U06755.1 | - | 4 - 5 | 614 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnn3
Genomic Map