Assay Details
| Gene Symbol: Mcoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039005.1 | NP_001034094.1 | 12 - 13 | 1591 | 74 | |
| GenBank mRNA | BC099787.1 | - | 12 - 13 | 1591 | 7934506 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P770336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55225 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P770336 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P770336 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P770336 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P335369 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43386 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10819703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819697 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10819690 | - | - |
| Rat Genome 230 2.0 Array | 1385587_at | - | - |
| Rat Genome 230 2.0 Array | 1385587_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385587_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385587_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200016626.rn.1 | TC0200002368.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200016625.rn.1 | TC0200002368.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385587_at | - | - |
| Rat Expression Set 230 | 1385587_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22413005 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362411 | - | - |

Mcoln2
Genomic Map